missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GORASP2 Mouse anti-Human, Porcine, Rat, Clone: 1C9A3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66627-1-IG-20UL
This item is not returnable.
View return policy
Description
GRASP55 is a PDZ containing protein involved in the stacking of golgi cisternea in a cell-free system.Specifications
| GORASP2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9H8Y8, Q9R064 | |
| Gorasp2 | |
| GORASP2 Fusion Protein Ag17607 | |
| 20 μL | |
| Primary | |
| Rat, Pig, Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry | |
| 1C9A3 | |
| Unconjugated | |
| GORASP2 | |
| Golgi phosphoprotein 6, GOLPH6, GORASP2, GRASP55, GRS2, p59 | |
| Mouse | |
| Protein G | |
| RUO | |
| 100154960, 113961, 26003 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction