missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GORASP2 Mouse anti-Human, Porcine, Rat, Clone: 1C9A3, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | GORASP2 |
|---|---|
| Clone | 1C9A3 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16808685
|
Proteintech
66627-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16898675
|
Proteintech
66627-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GRASP55 is a PDZ containing protein involved in the stacking of golgi cisternea in a cell-free system.Specifications
| GORASP2 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Golgi phosphoprotein 6, GOLPH6, GORASP2, GRASP55, GRS2, p59 | |
| Gorasp2 | |
| IgG1 | |
| Protein G | |
| Antibody |
| 1C9A3 | |
| Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Rat, Pig, Human | |
| Q9H8Y8, Q9R064 | |
| 100154960, 113961, 26003 | |
| GORASP2 Fusion Protein Ag17607 | |
| Primary | |
| -20°C | |
| GORASP2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title