missing translation for 'onlineSavingsMsg'
Learn More

ThermoScientific™ GoPure™ AAVX Affinity Membrane, 1 mL

Product Code. 31662815 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 Device

Product Code. 31662815

Brand: ThermoScientific™ A40003668

Please to purchase this item. Need a web account? Register with us today!

The GoPure AAVX Affinity Membrane provides efficient, small-scale capture of AAV viral vectors for screening of engineered capsids, as well as analytical sample preparation.

The GoPure AAVX Affinity Membrane is designed to provide rapid capture of adeno-associated virus (AAV) vectors at small scale. Incorporating CaptureSelect AAVX ligand, the membrane chromatography unit demonstrates broad serotype specificity to both naturally occurring and synthetic AAV capsids.

Features of the GoPure AAVX Affinity Membrane include:

  • Up to 5–15 times faster processing time than resin chromatography*
  • Broad AAV specificity, including AAV1–AAV8, AAVrh10, and synthetic serotypes
  • Binds >1 x 1014 capsids at 10 second residence time
  • High yield of AAV viral vectors directly from cell culture lysates

The GoPure AAVX Affinity Membrane provides efficient, small-scale capture of AAV viral vectors for screening of engineered capsids, as well as analytical sample preparation.

Main characteristics

Membrane bed: proprietary ePTFE composite

Nominal bed volume: 1 mL

Ligand: single-domain [VHH] antibody fragment

Binding capacity: >1 x 1014 cp/mL at 10 second residence time

Maximum operating pressure: 0.50 MPa (5.0 bar, 72.5 psi)

Formulation buffer: 18% (v/v) ethanol

*Users can typically purify up to 5–15 times faster compared to AAVX Affinity Resin at three-minute residence time.

Specifications

Type Affinity Matrix
Quantity 1 Device
Binding Capacity >1 x 1014 cp/mL
Ligand Type CaptureSelect™ AAVX
Matrix Proprietary ePTFE composite
Volume (Metric) 1 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.