missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GOLM1 Rabbit anti-Human, Clone: 15, Novus Biologicals™
Rabbit Monoclonal Antibody
£286.00 - £429.00
Specifications
| Antigen | GOLM1 |
|---|---|
| Clone | 15 |
| Dilution | Flow Cytometry 1:25-1:100, Immunocytochemistry/Immunofluorescence |
| Applications | Flow Cytometry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18254727
|
Novus Biologicals
NBP2-90164-50UL |
50 μg |
£286.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18260027
|
Novus Biologicals
NBP2-90164-100UL |
100 μg |
£429.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GOLM1 Monoclonal specifically detects GOLM1 in Human samples. It is validated for Flow Cytometry.Specifications
| GOLM1 | |
| Flow Cytometry 1:25-1:100, Immunocytochemistry/Immunofluorescence | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| bA379P1.3, FLJ23608, golgi membrane protein 1, Golgi membrane protein GP73, Golgi phosphoprotein 2C9orf155, golgi protein, 73-kD, GOLPH2 FLJ22634, GP73, GP73 chromosome 9 open reading frame 155, PSEC0257 | |
| Recombinant Human GOLM1 protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 15 | |
| Flow Cytometry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Golgi Apparatus Markers, Signal Transduction | |
| 0.2 um filtered solution in PBS | |
| 51280 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title