missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GOLM1 Rabbit anti-Human, Clone: 15, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90164-100UL
This item is not returnable.
View return policy
Description
GOLM1 Monoclonal specifically detects GOLM1 in Human samples. It is validated for Flow Cytometry.
Specifications
| GOLM1 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunofluorescence | |
| 15 | |
| Flow Cytometry 1:25-1:100, Immunocytochemistry/Immunofluorescence | |
| bA379P1.3, FLJ23608, golgi membrane protein 1, Golgi membrane protein GP73, Golgi phosphoprotein 2C9orf155, golgi protein, 73-kD, GOLPH2 FLJ22634, GP73, GP73 chromosome 9 open reading frame 155, PSEC0257 | |
| Recombinant Human GOLM1 protein | |
| 100 μg | |
| Golgi Apparatus Markers, Signal Transduction | |
| 51280 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction