missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GOLM1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£147.00 - £376.00
Specifications
| Antigen | GOLM1 |
|---|---|
| Concentration | 0.61 mg/mL |
| Dilution | Western Blot 1:5000, Immunocytochemistry/Immunofluorescence 1:500, Knockout Validated |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence, KnockDown |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18271104
|
Novus Biologicals
NBP1-50627SS |
0.025 mL |
£147.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18749833
|
Novus Biologicals
NBP1-50627 |
0.1 mL |
£376.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GOLM1 Polyclonal specifically detects GOLM1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Knockout Validated.Specifications
| GOLM1 | |
| Western Blot 1:5000, Immunocytochemistry/Immunofluorescence 1:500, Knockout Validated | |
| Polyclonal | |
| Rabbit | |
| Golgi Apparatus Markers, Signal Transduction | |
| PBS, 0.1% BSA, and 50% Glycerol with 0.05% Sodium Azide | |
| bA379P1.3, FLJ23608, golgi membrane protein 1, Golgi membrane protein GP73, Golgi phosphoprotein 2C9orf155, golgi protein, 73-kD, GOLPH2 FLJ22634, GP73, GP73 chromosome 9 open reading frame 155, PSEC0257 | |
| GOLM1 | |
| IgG | |
| Affinity Purified |
| 0.61 mg/mL | |
| Western Blot, Immunocytochemistry, Immunofluorescence, KnockDown | |
| Unconjugated | |
| RUO | |
| Human, Mouse | |
| Q8NBJ4 | |
| 51280 | |
| A genomic peptide made to an internal region of the human GOLM1 protein (within residues 250-400). [Swiss-Prot Q8NBJ4] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title