missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GOLM1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-50627
This item is not returnable.
View return policy
Description
GOLM1 Polyclonal specifically detects GOLM1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Knockout Validated.
Specifications
| GOLM1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 0.1% BSA, and 50% Glycerol with 0.05% Sodium Azide | |
| bA379P1.3, FLJ23608, golgi membrane protein 1, Golgi membrane protein GP73, Golgi phosphoprotein 2C9orf155, golgi protein, 73-kD, GOLPH2 FLJ22634, GP73, GP73 chromosome 9 open reading frame 155, PSEC0257 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence, KnockDown | |
| 0.61 mg/mL | |
| Western Blot 1:5000, Immunocytochemistry/Immunofluorescence 1:500, Knockout Validated | |
| Q8NBJ4 | |
| GOLM1 | |
| A genomic peptide made to an internal region of the human GOLM1 protein (within residues 250-400). [Swiss-Prot Q8NBJ4] | |
| 0.1 mL | |
| Golgi Apparatus Markers, Signal Transduction | |
| 51280 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction