missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GOLGA5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-60047
This item is not returnable.
View return policy
Description
GOLGA5 Polyclonal specifically detects GOLGA5 in Human samples. It is validated for Western Blot.
Specifications
| GOLGA5 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| Cell proliferation-inducing gene 31 protein, golgi autoantigen, golgin subfamily a, 5, golgin A5, Golgin subfamily A member 5, Golgin-84, GOLIM5, Protein Ret-II, PTC5, RET-fused gene 5 protein, ret-II, RETII, rfg5, RFG5golgi integral membrane protein 5 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Expected identity based on immunogen sequence: Rat: 100%; Bovine: 92%; Canine: 92%; Equine: 92%; Pig: 92%; Chicken: 76%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q8TBA6 | |
| GOLGA5 | |
| Synthetic peptides corresponding to GOLGA5(golgi autoantigen, golgin subfamily a, 5) The peptide sequence was selected from the N terminal of GOLGA5. Peptide sequence FVRRKKSEPDDELLFDFLNSSQKEPTGRVEIRKEKGKTPVFQSSQTSSVS. | |
| 100 μL | |
| Golgi Apparatus Markers | |
| 9950 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction