Learn More
3M™ GoggleGear™ Protective Goggle 3000 Series, PVC Shroud, Clear Scotchgard™ AF Lens, Premium Cloth Strap
Product Code. 31875402 Shop All 3M Products
Change view
Click to view available options
Frame Material:
PVC
Unit Size:
Each
Product Code. Frame Material Unit Size
31875402 PVC Each
1 options
Product Code. 31875402 Supplier 3M™ Supplier No. 7100325012
  • ONLINE EXCLUSIVE PRICE
    £19.85
    £12.27 / Pack of 1
    Save £7.58
    38% Off
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Protective goggle 3000 series features a full-size design with wraparound lens for improved field of vision, and slotted nose bridge for a more comfortable fit
  • 3M™ GoggleGear™ Protective Goggle 3000 Series features a full-size design with wrap-around lens for improved field of vision, and slotted nose bridge for a more comfortable fit.
  • Choose from models with anti-fog or 3M™ Scotchgard™ Anti-Fog coating.
  • The GoggleGear™ 3000 is available with indirect venting for enhanced air circulation and anti-fog performance, and is 3, protection against droplets, 4, protection against large particles, and 6, protection against streams of liquid, rated.
  • Includes either an adjustable polyester cloth strap, or easy to clean adjustable neoprene strap, as well as an optimized shroud design with slots to accommodate many prescription eyewear.
  • Acetate lens model meets the chemical resistance requirement of ISO 16321, helping protect workers from chemical hazards.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog
Color Clear
Lens Tint Clear
Product Type Anti-fog Goggles
Disposable Reusable
Lens Material Polycarbonate
Frame Material PVC
UV Protection None
Frame Type Full Frame