Learn More
3M™ Goggle Gear™ Safety Goggles, Anti-Fog / Anti-Scratch Coating (K&N)
Product Code. 17227680
Change view
Click to view available options
Lens Tint:
Gray
Transparent
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Lens Tint unitSize
17227680 Transparent Pack of 10
17237680 Gray Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17227680 Supplier 3M™ Supplier No. 7100185183
Add to basket
Add to basket
Goggle Gear™ Anti-Fog / Anti-Scratch Coating (K&N)
  • Modern slim-line and lightweight design for an excellent fit
  • Adjustable Neoprene headband for easy cleaning and sterilisation
  • Pivoting ratchet for convenient personal adjustment
  • 3M™ Scotchgard™ Anti-Fog Coating provides superior anti-fog and anti-scratch properties
  • Coating withstands washing, soaking in bleach or using alcohol wipes without losing performance
  • Special 3M™ Goggle Gear prescription insert available
  • Offers excellent protection against UV radiation
  • Lens Marking: 2C-1.2 3M 1 BT KN
  • Frame Marking: 3M EN 166 3 4 BT CE
  • Protection against large dust particles (3)
  • Protection against liquid droplets (4)
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog
Lens Tint Transparent
Frame Color Gray and Red
Model GG501NSGAF-EU
Quantity 10/Cs.
Lens Material Polycarbonate
Frame Type Goggle