missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Goat anti-Mouse IgG1 Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 633

Product Code. 10052642
Change view
Click to view available options
Quantity:
500 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10052642 500 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10052642 Supplier Invitrogen™ Supplier No. A21126

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Goat Polyclonal Secondary Antibody

Fluorescence of this long-wavelength Alexa Fluor dye is not visible by looking through a conventional fluorescence microscope. Product will be shipped at Room Temperature.

Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse IgG1 Cross-Adsorbed
Applications Flow Cytometry, Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Alexa Fluor 633
Formulation PBS with 5mM sodium azide; pH 7.5
Host Species Goat
Immunogen IgG gamma 1.
Purification Method Purified
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Mouse
Content And Storage 4°C, store in dark
Product Type Secondary Antibody
Form Liquid
Isotype IgG
Show More Show Less
I have two primary antibodies from the same host species that I wish to use to label the same cells. How do I choose secondary antibodies?

If the two primary antibodies are from different isotypes (such as mouse IgG1 and IgG2a), there are isotype-specific secondary antibodies to choose from. The other option is to make direct conjugates of the primary antibodies through the use of antibody label kits, reactive dyes, or custom conjugations.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.