Learn More
Invitrogen™ Goat anti-Mouse IgG (H+L) Superclonal™ Secondary Antibody, Alexa Fluor™ 647
Description
The sensitivity and specificity of each lot is confirmed using Immunocytochemistry. Minimal cross-reactivity with rabbit, rat, human, bovine, guinea pig, and donkey IgG is observed. Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include: better specificity, animal origin-free formulation, and more lot-to-lot consistency.
Specifications
Specifications
| Antigen | Mouse IgG (H+L) |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Recombinant Superclonal |
| Concentration | 1 mg/mL |
| Conjugate | Alexa Fluor 647 |
| Formulation | PBS with 5mM sodium azide |
| Host Species | Goat |
| Immunogen | Recombinant full-length protein. |
| Purification Method | Gravity column chromatography |
| Quantity | 1 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.