Learn More
Invitrogen™ Goat anti-Human IgG Fc Cross-Adsorbed Secondary Antibody
Description
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31125 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 31125 reacts with the heavy chains of human IgG from only the Fc portion, but not with the light chains or the Fab fragment common to most human immunoglobulins. This antibody does not react against human IgM, IgA or non-immunoglobulin serum proteins. The antibody has been tested by ELISA and/or solid-phase adsorbed to ensure minimal cross-reaction with bovine, horse and mouse serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Store product at 4°C until opened. After opening, dilute only enough antibody for a single day's use. Store remainder at 4°C under sterile conditions. Country of origin: USA.
Specifications
Specifications
| Antigen | Human IgG Fc Cross-Adsorbed |
| Applications | Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.6 |
| Host Species | Goat |
| Purification Method | Antigen affinity chromatography |
| Quantity | 1 mg |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.