missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GnRH1 Monoclonal Antibody (LHRH13.327.8), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA5-46094
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
MA1-10814 detects Gonadotropin Releasing Hormone from human samples. MA1-10814 has been successfully used in ELISA, radioimmunoassays and immunohistochemistry applications. The MA1-10814 immunogen is purified human gonadotropin releasing hormone.
The Luteinizing Hormone Releasing Hormone (LH-RH) is produced in the hypothalamus and influences the release of Luteinizing Hormone in the pituitary gland. LH-RH is also known as Gonadotropin-releasing hormone (Gn-RH).
Tekniske data
| GnRH1 | |
| Monoclonal | |
| 0.76 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| P01148 | |
| GNRH1 | |
| Purified human gonadotropin releasing hormone. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Immunohistochemistry, Radioimmune Assays (RIA) | |
| LHRH13.327.8 | |
| Unconjugated | |
| GNRH1 | |
| Gnrh; Gnrh1; Gnrh2; Gnrha; GnRH-associated peptide 1; GnRH-associated peptide I; GnRH-I; Gonadoliberin I; Gonadoliberin-1; Gonadorelin; gonadotropin releasing hormone 1; gonadotropin releasing hormone 2; gonadotropin-releasing hormone 1 (luteinizing-releasing hormone); gonadotropin-releasing hormone 1 variant A; gonadotropin-releasing hormone 1 variant B; Gonadotropin-releasing hormone I; GRH; hpg; hypogonadal; leuteinizing-releasing hormone; LHRH; LH-RH I; Lhrh1; LNRH; Luliberin I; luteinizing hormone releasing hormone; luteinizing hormone-releasing hormone; luteinizing hormone-releasing hormone I; Lutropin; Progonadoliberin I; progonadoliberin-1; prolactin release-inhibiting factor; Prolactin release-inhibiting factor 1; Prolactin release-inhibiting factor I; Rgnrhg1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 2796 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion