missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GnRH1 Monoclonal Antibody (LHRH13.327.8), Invitrogen™

Mouse Monoclonal Antibody
£367.00 - £916.00
Specifications
| Antigen | GnRH1 |
|---|---|
| Clone | LHRH13.327.8 |
| Concentration | 0.76 mg/mL |
| Applications | ELISA, Immunohistochemistry, Radioimmune Assays (RIA) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17880406
|
Invitrogen™
MA5-46094 |
100 μg |
£367.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11563423
![]() |
Invitrogen™
MA110814 |
500 μg |
£506.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17810476
|
Invitrogen™
MA5-46173 |
1 mg |
£916.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MA1-10814 detects Gonadotropin Releasing Hormone from human samples. MA1-10814 has been successfully used in ELISA, radioimmunoassays and immunohistochemistry applications. The MA1-10814 immunogen is purified human gonadotropin releasing hormone.
The Luteinizing Hormone Releasing Hormone (LH-RH) is produced in the hypothalamus and influences the release of Luteinizing Hormone in the pituitary gland. LH-RH is also known as Gonadotropin-releasing hormone (Gn-RH).Specifications
| GnRH1 | |
| 0.76 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative; pH 7.4 | |
| Gnrh; Gnrh1; Gnrh2; Gnrha; GnRH-associated peptide 1; GnRH-associated peptide I; GnRH-I; Gonadoliberin I; Gonadoliberin-1; Gonadorelin; gonadotropin releasing hormone 1; gonadotropin releasing hormone 2; gonadotropin-releasing hormone 1 (luteinizing-releasing hormone); gonadotropin-releasing hormone 1 variant A; gonadotropin-releasing hormone 1 variant B; Gonadotropin-releasing hormone I; GRH; hpg; hypogonadal; leuteinizing-releasing hormone; LHRH; LH-RH I; Lhrh1; LNRH; Luliberin I; luteinizing hormone releasing hormone; luteinizing hormone-releasing hormone; luteinizing hormone-releasing hormone I; Lutropin; Progonadoliberin I; progonadoliberin-1; prolactin release-inhibiting factor; Prolactin release-inhibiting factor 1; Prolactin release-inhibiting factor I; Rgnrhg1 | |
| GNRH1 | |
| IgG1 | |
| Protein G | |
| Antibody |
| LHRH13.327.8 | |
| ELISA, Immunohistochemistry, Radioimmune Assays (RIA) | |
| Unconjugated | |
| Mouse | |
| Human | |
| P01148 | |
| 2796 | |
| Purified human gonadotropin releasing hormone. | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| GNRH1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title