missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GNE Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25079-1-AP-20UL
This item is not returnable.
View return policy
Description
GNE is a bifunctional enzyme that initiates and regulates the biosynthesis of N-acetylneuraminic acid (NeuAc), a precursor of sialic acids. It is a rate-limiting enzyme in the sialic acid biosynthetic pathway. Sialic acid modification of cell surface molecules is crucial for their function in many biologic processes, including cell adhesion and signal transduction. Differential sialylation of cell surface molecules is also implicated in the tumorigenicity and metastatic behavior of maligt cells.Specifications
| GNE | |
| Polyclonal | |
| Unconjugated | |
| GNE | |
| DMRV, GLCNE, GNE, IBM2, ManAc kinase, NM, Uae1, UDP GlcNAc 2 epimerase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10020 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
| 0.39 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9Y223 | |
| GNE | |
| GNE Fusion Protein Ag18689 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction