missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GNAO1 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
£119.00 - £295.00
Specifications
| Antigen | GNAO1 |
|---|---|
| Clone | 1B5G2 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30235281
|
Proteintech
68027-1-IG-150UL |
150 μL |
£349.00 £295.00 / 150µL Save £54.00 15% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30237136
|
Proteintech
68027-1-IG-20UL |
20 μL |
£135.00 £119.00 / 20µL Save £16.00 11% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The GNAO1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GNAO1. This antibody recognizes mouse, rat, rabbit, pig, chicken, human antigen. The GNAO1 antibody has been validated for the following applications: WB, IHC, IF-P, ELISA analysis.Specifications
| GNAO1 | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 2775 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 1B5G2 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| Unconjugated | |
| Mouse | |
| Mouse, Rat, Rabbit, Pig, Chicken, Human | |
| DKFZp686O0962; G-ALPHA-o; GNAO | |
| IgG1 | |
| Protein G purification | |
| Monoclonal Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title