missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GNAO1 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68027-1-IG-150UL
This item is not returnable.
View return policy
Description
The GNAO1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GNAO1. This antibody recognizes mouse, rat, rabbit, pig, chicken, human antigen. The GNAO1 antibody has been validated for the following applications: WB, IHC, IF-P, ELISA analysis.Specifications
| GNAO1 | |
| Monoclonal | |
| 1000 μg/mL | |
| DKFZp686O0962; G-ALPHA-o; GNAO | |
| Recombinant Protein | |
| 150 μL | |
| Primary | |
| Mouse, Rat, Rabbit, Pig, Chicken, Human | |
| 40 kDa | |
| Liquid |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| 1B5G2 | |
| Unconjugated | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 2775 | |
| Store at -20°C. Stable for one year after shipment. | |
| Monoclonal Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction