missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GNAL Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-68695
This item is not returnable.
View return policy
Description
GNAL Polyclonal antibody specifically detects GNAL in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| GNAL | |
| Polyclonal | |
| Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Adenylate cyclase-stimulating G alpha protein, olfactory type, guanine nucleotide binding protein (G protein), alpha activating activitypolypeptide, olfactory typ, guanine nucleotide binding protein (G protein), alpha activating activitypolypeptide, olfactory type, guanine nucleotide binding protein (G protein), alpha stimulating activitypolypeptide, olfactory type, guanine nucleotide-binding protein G(olf) subunit alpha | |
| This antibody was developed against a recombinant protein corresponding to amino acids: NQFRSDYIKSIAPITDFEYSQEFFDHVKKLWDDEGVKACFER | |
| 100 μg | |
| Signal Transduction | |
| 2774 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction