missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GNA11 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00 - £513.00
Specifications
| Antigen | GNA11 |
|---|---|
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18187389
|
Novus Biologicals
NBP2-54735 |
100 μL |
£513.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18650918
|
Novus Biologicals
NBP2-54735-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GNA11 Polyclonal specifically detects GNA11 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| GNA11 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| G alpha-11, GA11, GNA-11, G-protein subunit alpha-11, guanine nucleotide binding protein (G protein), alpha 11 (Gq class), Guanine nucleotide-binding protein G(y) subunit alpha, guanine nucleotide-binding protein subunit alpha-11, guanine nucleotide-binding protein, Gq class, GNA11 | |
| GNA11 | |
| IgG | |
| Immunogen affinity purified |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 2767 | |
| This antibody was developed against a Recombinant Protein corresponding to amino acids:SDNENRMEESKALFRTIITYPWFQNS | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title