missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GNA11 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-54735
This item is not returnable.
View return policy
Description
GNA11 Polyclonal specifically detects GNA11 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| GNA11 | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| G alpha-11, GA11, GNA-11, G-protein subunit alpha-11, guanine nucleotide binding protein (G protein), alpha 11 (Gq class), Guanine nucleotide-binding protein G(y) subunit alpha, guanine nucleotide-binding protein subunit alpha-11, guanine nucleotide-binding protein, Gq class, GNA11 | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 2767 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| GNA11 | |
| This antibody was developed against a Recombinant Protein corresponding to amino acids:SDNENRMEESKALFRTIITYPWFQNS | |
| 100 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion