Learn More
Pall GN-4 Metricel™ Membrane Disc Filters
Certified 0.8 μm membrane for microbiological analysis meets or surpasses regulatory specifications.
Brand:  Pall 66179
Additional Details : Weight : 0.25650kg
 View more versions of this product
Product Code. 17144551
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Designed for laboratory applications only
  • Not approved for use in medical, clinical, surgical or other patient applications
Specifications
Specifications
Hydrophilic MCE
Non-sterile
152 μm
47 mm
0.8 μm
100/Pk.
Gridded
Sugerencias de productos
Sugerencias de productos
Videos
SDS
Documentos
Documentos
Product Certifications
Ofertas especiales
Ofertas especiales