Learn More
Pall GN-4 Metricel™ Membrane Disc Filters
Certified 0.8 μm membrane for microbiological analysis meets or surpasses regulatory specifications.
Brand: Pall 64679
Additional Details : Weight : 0.00900kg Transport : UN number : 3270 Chem class : 4.1 Pack group : II
Description
- Designed for laboratory applications only
- Not approved for use in medical, clinical, surgical or other patient applications
Specifications
Hydrophilic MCE | |
Non-sterile | |
152 μm | |
47 mm |
0.8 μm | |
100/Pk. | |
Plain |
Safety and Handling
- Membranes: cellulose nitrate and esters of cellulose
Signal Word
- Danger
Hazard Category
- Flammable solid Category 1
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
- MIXTURE LIST-Contains : nitrocellulose