Learn More
GMF Beta Mouse, anti-Human, CoraLite Plus 488, Clone: 3B10A2, Proteintech
Mouse Monoclonal Antibody
£135.00 - £375.00
Specifications
| Antigen | GMF Beta |
|---|---|
| Clone | 3B10A2 |
| Concentration | 1000 μg/mL |
| Applications | Immunofluorescence |
| Conjugate | CoraLite Plus 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18099645
|
Proteintech
CL488-60062-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18079825
|
Proteintech
CL488-60062-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The GMF Beta antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GMF Beta. This antibody recognizes human, pig antigen. The GMF Beta antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| GMF Beta | |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Mouse | |
| 2764 | |
| IgG1 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 3B10A2 | |
| Immunofluorescence | |
| Liquid | |
| Human, Pig | |
| Glia maturation factor beta, glia maturation factor, beta, GMF, GMF beta, GMFB | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.