Learn More
GMF Beta Mouse, anti-Human, CoraLite Plus 488, Clone: 3B10A2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-60062-20UL
Description
The GMF Beta antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GMF Beta. This antibody recognizes human, pig antigen. The GMF Beta antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| GMF Beta | |
| 3B10A2 | |
| CoraLite Plus 488 | |
| Mouse | |
| Primary | |
| Human, Pig | |
| 155 kDa | |
| IgG1 |
| Immunofluorescence | |
| 1000 μg/mL | |
| Glia maturation factor beta, glia maturation factor, beta, GMF, GMF beta, GMFB | |
| 20 μL | |
| 2764 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |