missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GlyT2 Recombinant Rabbit Monoclonal Antibody (23GB4145), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552663
This item is not returnable.
View return policy
Description
This gene encodes a sodium- and chloride-dependent glycine neurotransmitter transporter. This integral membrane glycoprotein is responsible for the clearance of extracellular glycine during glycine-mediated neurotransmission.
Specifications
| GlyT2 | |
| Recombinant Monoclonal | |
| 1.2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P58295, Q761V0, Q9Y345 | |
| SLC6A5 | |
| A synthesized peptide derived from human Glyt2 (301-336AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB4145 | |
| Unconjugated | |
| SLC6A5 | |
| glycine transporter 2; GLYT2; glyT-2; HKPX3; NET1; prestin; si:dkey-7n10.12; slc6a5; Sodium- and chloride-dependent glycine transporter 2; solute carrier family 6 (neurotransmitter transporter), member 5; solute carrier family 6 (neurotransmitter transporter, glycine), member 5; solute carrier family 6 member 5 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 104245, 171148, 9152 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction