missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GlyT2 Recombinant Rabbit Monoclonal Antibody (23GB4145), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | GlyT2 |
|---|---|
| Clone | 23GB4145 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089671
![]() |
Invitrogen™
MA552662 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089298
|
Invitrogen™
MA552663 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GlyT2 Recombinant Monoclonal antibody specifically detects GlyT2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| GlyT2 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P58295, Q761V0, Q9Y345 | |
| 104245, 171148, 9152 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB4145 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| glycine transporter 2; GLYT2; glyT-2; HKPX3; NET1; prestin; si:dkey-7n10.12; slc6a5; Sodium- and chloride-dependent glycine transporter 2; solute carrier family 6 (neurotransmitter transporter), member 5; solute carrier family 6 (neurotransmitter transporter, glycine), member 5; solute carrier family 6 member 5 | |
| A synthesized peptide derived from human Glyt2 (301-336AA). | |
| Primary | |
| -20°C | |
| SLC6A5 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title