missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glyoxalase I Antibody (9F11), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59369
This item is not returnable.
View return policy
Description
Glyoxalase I Monoclonal specifically detects Glyoxalase I in Non-species specific samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Glyoxalase I | |
| Monoclonal | |
| Unconjugated | |
| PBS pH 7.4, 50% glycerol with 0.09% Sodium Azide | |
| GLO1 | |
| Synthetic Methylglyoxal modified Keyhole Limpet Hemocyanin (KLH). | |
| 100 μg | |
| Cancer, Core ESC Like Genes, Signal Transduction, Stem Cell Markers | |
| 2739 | |
| Non-species specific | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| 9F11 | |
| Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
| Aldoketomutase, EC 4.4.1.5, GLOD1, Glx I, GLYI, glyoxalase domain containing 1, glyoxalase Ialdoketomutase, Ketone-aldehyde mutase, lactoyl glutathione lyase, lactoylglutathione lyase, methylglyoxalase, S-D-lactoylglutathione methylglyoxal lyase | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Specific for Methylglyoxal modified proteins. Does not detect free Methylglyoxal. Does not cross-react with Acrolein, Hexanoyl Lysine, Malondialdeyhde, 4-Hydroxy-2-hexenal, 4-Hydroxy nonenal, or Crotonaldehyde modified proteins. | |
| Store at -20C. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction