missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glyoxalase I Antibody (9F11), Novus Biologicals™
Mouse Monoclonal Antibody
£144.00 - £466.00
Specifications
| Antigen | Glyoxalase I |
|---|---|
| Clone | 9F11 |
| Dilution | Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18275382
|
Novus Biologicals
NBP2-59369 |
100 μg |
£466.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18610229
|
Novus Biologicals
NBP2-59369-25ug |
25 μg |
£144.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Glyoxalase I Monoclonal specifically detects Glyoxalase I in Non-species specific samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| Glyoxalase I | |
| Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Aldoketomutase, EC 4.4.1.5, GLOD1, Glx I, GLYI, glyoxalase domain containing 1, glyoxalase Ialdoketomutase, Ketone-aldehyde mutase, lactoyl glutathione lyase, lactoylglutathione lyase, methylglyoxalase, S-D-lactoylglutathione methylglyoxal lyase | |
| GLO1 | |
| IgG1 | |
| Protein G purified |
| 9F11 | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Cancer, Core ESC Like Genes, Signal Transduction, Stem Cell Markers | |
| 2739 | |
| Synthetic Methylglyoxal modified Keyhole Limpet Hemocyanin (KLH). | |
| Primary | |
| Specific for Methylglyoxal modified proteins. Does not detect free Methylglyoxal. Does not cross-react with Acrolein, Hexanoyl Lysine, Malondialdeyhde, 4-Hydroxy-2-hexenal, 4-Hydroxy nonenal, or Crotonaldehyde modified proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title