missing translation for 'onlineSavingsMsg'
Learn More

Glyoxal sodium bisulfite hydrate, 98%, Thermo Scientific™

Product Code. p-7077178 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
250 g
1 kg
Unit Size:
1kg
250g
25g
This item is not returnable. View return policy

Product Code. 15577029

Brand: Thermo Scientific Acros 173270250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 332360-05-1
Molecular Formula C2H4Na2O8S2
Molecular Weight (g/mol) 266.15
MDL Number MFCD0149541
InChI Key BXUKAXFDABMVND-UHFFFAOYNA-L
Synonym glyoxal-sodium bisulfite monohydrate, glyoxal sodium bisulfite addition compound hydrate, 1,2-dihydroxy-1,2-ethanedisulfonic acid disodium salt hydrate, 1,2-dihydroxy-1,2-ethanedisulfonic acid disodium salt, c2h4o8s2.2na.h2o, glyoxal-sodium bisulfite monohydrate mi, glyoxal bis sodium hydrogen sulfite adduct hydrate, glyoxal sodium bisulfite addition compound hydrate,98, disodium 1,2-dihydroxy-1,2-ethanedisulfonate monohydrate, disodium 1,2-dihydroxyethane-1,2-disulfonate hydrate
PubChem CID 16211366
SMILES [Na+].[Na+].OC(C(O)S([O-])(=O)=O)S([O-])(=O)=O

Specifications

Melting Point 193.0°C to 195.0°C
Quantity 25 g
Assay Percent Range 97.5% min
Linear Formula [-CH(OH)SO3Na]2·xH2O
Merck Index 14,451
Solubility Information Solubility in water: freely soluble. Other solubilities: practically insoluble in alcohol
IUPAC Name disodium 1,2-dihydroxyethane-1,2-disulfonate
Formula Weight 266.16
Percent Purity 98%
Infrared Spectrum Authentic
Loss on Drying 10% max. (120°C, 2 hrs)
Chemical Name or Material Glyoxal sodium bisulfite hydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.