accessibility menu, dialog, popup

UserName

CAS RN 332360-05-1

SSOOOOOOOHOHNaNa

CAS RN 332360-05-1

IUPAC Name: (1R,2R)-1,2-dihydroxyethane-1,2-disulfonate
Synonyms: (1R,2R)-1,2-dihydroxyethane-1,2-disulfonate
Molecular Weight (g/mol): 220.17
Molecular Formula: C2H4O8S2
InChi Key: HADMFXUIQRIRMV-JCYAYHJZSA-L
SMILES: O[C@@H]([C@H](O)S([O-])(=O)=O)S([O-])(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results