missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glyoxal sodium bisulfite hydrate, 98%, Thermo Scientific™
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
250 g
1 kg
Unit Size:
1kg
250g
25g
Specifications
Specifications
| Melting Point | 193.0°C to 195.0°C |
| Quantity | 250 g |
| Assay Percent Range | 97.5% min |
| Linear Formula | [-CH(OH)SO3Na]2·xH2O |
| Merck Index | 14,451 |
| Solubility Information | Solubility in water: freely soluble. Other solubilities: practically insoluble in alcohol |
| IUPAC Name | disodium;1,2-dihydroxyethane-1,2-disulfonate;hydrate |
| Formula Weight | 266.16 |
| Percent Purity | 98% |
| Infrared Spectrum | Authentic |
| Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction