missing translation for 'onlineSavingsMsg'
Learn More

Glyme (1,2-dimethoxyethane-d10), TRC

Product Code. 30240808
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30240808 100 mg 100mg
30240743 1 g 1g
30240772 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30240808 Supplier Toronto Research Chemicals Supplier No. TRCG752003100MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107975-86-0
Molecular Formula C4D10O2
Molecular Weight (g/mol) 100.18
Synonym 1,2-Dimethoxyethane D10
IUPAC Name 1,1,2,2-tetradeuterio-1,2-bis(trideuteriomethoxy)ethane
SMILES [2H]C([2H])([2H])OC([2H])([2H])C([2H])([2H])OC([2H])([2H])[2H]

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C4H10O2/c1-5-3-4-6-2/h3-4H2,1-2H3/i1D3,2D3,3D2,4D2
Formula Weight 100.1308
Chemical Name or Material Glyme (1,2-dimethoxyethane-d10)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.