missing translation for 'onlineSavingsMsg'
Learn More

Glyme (1,2-dimethoxyethane-d10), TRC

Product Code. 30240743
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30240743 1 g 1g
30240772 500 mg 500mg
30240808 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30240743 Supplier Toronto Research Chemicals Supplier No. TRCG7520031G
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107975-86-0
Molecular Formula C4D10O2
Molecular Weight (g/mol) 100.18
Synonym 1,2-Dimethoxyethane D10
IUPAC Name 1,1,2,2-tetradeuterio-1,2-bis(trideuteriomethoxy)ethane
SMILES [2H]C([2H])([2H])OC([2H])([2H])C([2H])([2H])OC([2H])([2H])[2H]

Specifications

Quantity 1 g
InChI Formula InChI=1S/C4H10O2/c1-5-3-4-6-2/h3-4H2,1-2H3/i1D3,2D3,3D2,4D2
Formula Weight 100.1308
Chemical Name or Material Glyme (1,2-dimethoxyethane-d10)
Product Title
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.