Learn More
Thermo Scientific™ GlycoLink™ IP Kit
Shop All Thermo Scientific ProductsDescription
The Thermo Scientific GlycoLink IP Kit provides components for effective immunoprecipitation (IP and co-IP) based on small-scale, covalent, affinity-resin immobilization of antibodies and other glycoproteins via oxidized sugar groups.
Highlights:
Efficient immobilization – couple 2 to 10μg of oxidized antibody or other glycoprotein to 20μL of resin in 2 hours or less
Stable immobilization – resonance structure of the hydrazone bonds are sufficiently stable so the antibody remains coupled during elution
Preserves binding function – immobilizes IgG via the Fc region, thereby keeping both antigen binding sites available for capturing target
Versatile applications – immobilize any molecule that contains oxidizable sugars, including glycoproteins for use in purifying protein interaction binding partners
Includes:
Kit contains resin, buffers and reagents for immobilization; desalting columns; IP lysis/wash and elution buffers for immunoprecipitation; microcentrifuge spin columns and collection tubes; and loading buffer for SDS-PAGE
Recommended for:
Immunoprecipitation followed by electrophoresis and excision of the protein for mass spectrometry or sequencing; Immunoprecipitation and SDS-PAGE analysis of a protein with a molecular weight similar to the antibody's heavy or light chain; Identification of binding partners of immobilized glycoprotein
Co-immunoprecipitation for determining protein-protein interactions
Specifications
Specifications
| Capacity (Metric) | 2 to 10 μg Oxidized Antibody or Glycoprotein Coupling to 20 μL Resin |
| Content And Storage | Sufficient For: 25 IP/Co-IP reactions, each using 20 μL antibody-coupled resin • UltraLink Hydrazide Resin, 0.5 mL • GlycoLink Coupling Buffer, 50 mL • Aniline, 0.2 mL • Sodium meta-periodate, 0.5 g • Wash Buffer, 60 mL • Zeba Spin Desalting Columns,0.5 mL, 25 columns • IP Lysis/Wash Buffer, 50 mL • PBS Pack (makes 500 mL), 1 pack • Elution Buffer, pH 2.8, 50 mL • Lane Marker Sample Buffer, Non-reducing, (5X), 5 mL • Pierce Spin Columns-Screw Caps, 25 columns • Collection Tubes, 2 mL, 100 tubes Store at 4°C. |
| Assay | Affinity Purification |
| For Use With (Equipment) | Microcentrifuge |
| Product Line | GlycoLink™ |
| Quantity | 25 reactions kit |
| For Use With (Application) | Immunoprecipitation (IP) |
| Target | Not Target-Specific |
| Type | Immunoprecipitation/Co-Immunoprecipitation Kit |
| Sufficient For | 25 Reactions |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.