missing translation for 'onlineSavingsMsg'
Learn More

Glycine-13C2, TRC

Product Code. 30391176
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30391176 100 mg
30397840 50 mg
30403333 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30391176 Supplier Toronto Research Chemicals Supplier No. TRCG615998100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67836-01-5
Molecular Formula 13C2H5NO2
Molecular Weight (g/mol) 77.05
Synonym 2-Aminoacetic-13C2 Acid, Aciport-13C2, Aminoethanoic-13C2 Acid, Glicoamin-13C2, Glycocoll-13C2, Glycolixir-13C2, Glycosthene-13C2, Gyn-Hydralin-13C2, H 1-13C2, NSC 25936-13C2, NSC 2916-13C2, NSC 54188-13C2, Padil-13C2
SMILES O[13C]([13C]([H])([H])N)=O

Specifications

InChI Formula InChI=1S/C2H5NO2/c3-1-2(4)5/h1,3H2,(H,4,5)/i1+1,2+1
Quantity 100 mg
Formula Weight 77.0387
Shipping Condition Room Temperature
Chemical Name or Material Glycine-13C2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.