CAS RN 67836-01-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Glycine-13C2, TRC
CAS: 67836-01-5 Molecular Formula: 13C2H5NO2 Molecular Weight (g/mol): 77.05 Synonym: 2-Aminoacetic-13C2 Acid,Aciport-13C2,Aminoethanoic-13C2 Acid,Glicoamin-13C2,Glycocoll-13C2,Glycolixir-13C2,Glycosthene-13C2,Gyn-Hydralin-13C2,H 1-13C2,NSC 25936-13C2,NSC 2916-13C2,NSC 54188-13C2,Padil-13C2 SMILES: O[13C]([13C]([H])([H])N)=O