missing translation for 'onlineSavingsMsg'
Learn More

Glycidyltrimethylammonium Chloride (Technical Grade), TRC

Product Code. 30392982
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30392982 100 mg -
30393650 1 g -
30400306 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30392982 Supplier Toronto Research Chemicals Supplier No. TRCG616008100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3033-77-0
Molecular Formula C6H14NO.Cl
Molecular Weight (g/mol) 151.63
Synonym (2,3-Epoxypropyl)trimethylammonium chloride, N,N,N-trimethyloxiranemethanaminium chloride
IUPAC Name trimethyl(oxiran-2-ylmethyl)azanium;chloride
SMILES [Cl-].C[N+](C)(C)CC1CO1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C6H14NO.ClH/c1-7(2,3)4-6-5-8-6;/h6H,4-5H2,1-3H3;1H/q+1;/p-1
Formula Weight 151.0764
Chemical Name or Material Glycidyltrimethylammonium Chloride (Technical Grade)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.