CAS RN 3033-77-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Glycidyltrimethylammonium Chloride (Technical Grade), TRC
CAS: 3033-77-0 Molecular Formula: C6H14NO.Cl Molecular Weight (g/mol): 151.63 Synonym: (2,3-Epoxypropyl)trimethylammonium chloride,N,N,N-trimethyloxiranemethanaminium chloride IUPAC Name: trimethyl(oxiran-2-ylmethyl)azanium;chloride SMILES: [Cl-].C[N+](C)(C)CC1CO1