missing translation for 'onlineSavingsMsg'
Learn More

Glycidyl Acrylate (Stabilized with MEHQ), TRC

Product Code. 30402881
Change view
Click to view available options
Quantity:
1 g
2.5 g
5 g
Unit Size:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30402881 5 g -
30407137 1 g -
30404322 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30402881 Supplier Toronto Research Chemicals Supplier No. TRCG6501735G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-90-1
Molecular Formula C6H8O3
Molecular Weight (g/mol) 128.13
Synonym 2-Propenoic acid, oxiranylmethyl ester (9CI), Acrylic acid, 2,3-epoxypropyl ester (6CI, 8CI), Acrylic acid, glycidyl ester (5CI), (Oxiran-2-yl)methyl prop-2-enoate, (±)-Glycidyl acrylate, 2,3-Epoxypropyl acrylate, A 1045, Epoxypropyl acrylate, Glycidyl acrylate, Glycidyl propenoate, NSC 24151, NSC 67196, SR 378
IUPAC Name oxiran-2-ylmethyl prop-2-enoate
SMILES C=CC(=O)OCC1CO1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C6H8O3/c1-2-6(7)9-4-5-3-8-5/h2,5H,1,3-4H2
Formula Weight 128.0473
Chemical Name or Material Glycidyl Acrylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.