CAS RN 106-90-1
IUPAC Name:
(oxiran-2-yl)methyl prop-2-enoate
Synonyms:
glycidyl acrylate
Molecular Weight (g/mol):
128.13
Molecular Formula:
C6H8O3
InChi Key:
RPQRDASANLAFCM-UHFFFAOYNA-N
SMILES:
C=CC(=O)OCC1CO1
1
–
1
of
1
results
1
Glycidyl Acrylate (Stabilized with MEHQ), TRC
CAS: 106-90-1 Molecular Formula: C6H8O3 Molecular Weight (g/mol): 128.13 Synonym: 2-Propenoic acid, oxiranylmethyl ester (9CI),Acrylic acid, 2,3-epoxypropyl ester (6CI, 8CI),Acrylic acid, glycidyl ester (5CI),(Oxiran-2-yl)methyl prop-2-enoate,(±)-Glycidyl acrylate,2,3-Epoxypropyl acrylate,A 1045,Epoxypropyl acrylate,Glycidyl acrylate,Glycidyl propenoate,NSC 24151,NSC 67196,SR 378 IUPAC Name: oxiran-2-ylmethyl prop-2-enoate SMILES: C=CC(=O)OCC1CO1