missing translation for 'onlineSavingsMsg'
Learn More

Glycidyl 4-methoxyphenyl ether, 98%

Product Code. 11458647
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11458647 100 g 100g
11448647 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11458647 Supplier Thermo Scientific Alfa Aesar Supplier No. A19780.22

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2211-94-1 | C10H12O3 | 180.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2211-94-1
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.20
MDL Number MFCD00005135
InChI Key AVWGFHZLPMLKBL-UHFFFAOYNA-N
Synonym glycidyl 4-methoxyphenyl ether, 2-4-methoxyphenoxy methyl oxirane, 2,3-epoxypropyl-4-methoxyphenyl ether, p-methoxyphenyl glycidyl ether, methoxyphenyl glycidyl ether, glycidyl p-methoxyphenyl ether, 2-p-methoxyphenoxymethyl oxirane, 4-methoxyphenoxy methyl oxirane, anisole, p-2,3-epoxypropoxy
PubChem CID 16646
IUPAC Name 2-[(4-methoxyphenoxy)methyl]oxirane
SMILES COC1=CC=C(OCC2CO2)C=C1

Specifications

Melting Point 45°C to 48°C
Flash Point >110°C (230°F)
Quantity 100 g
Formula Weight 180.2
Percent Purity 98%
Chemical Name or Material Glycidyl 4-methoxyphenyl ether

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.