accessibility menu, dialog, popup

UserName

CAS RN 2211-94-1

H3COOO

CAS RN 2211-94-1

IUPAC Name: 2-[(4-methoxyphenoxy)methyl]oxirane
Synonyms: 2-(4-methoxyphenoxymethyl)oxirane
Molecular Weight (g/mol): 180.2
Molecular Formula: C10H12O3
InChi Key: AVWGFHZLPMLKBL-UHFFFAOYNA-N
SMILES: COC1=CC=C(OCC2CO2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results