missing translation for 'onlineSavingsMsg'
Learn More

Glyceryl Tripalmitelaidate, TRC

Product Code. 30393571
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30393571 1 g
30403160 100 mg
30398535 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30393571 Supplier Toronto Research Chemicals Supplier No. TRCG6015551G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134907-85-0
Molecular Formula C51H92O6
Molecular Weight (g/mol) 801.27
Synonym (9E,9'E,9''E)-9-Hexadecenoic Acid 1,2,3-Propanetriyl Ester, (E,E,E)-9-Hexadecenoic Acid 1,2,3-Propanetriyl Ester, Tripalmitelaidin
IUPAC Name 2,3-bis[[(E)-hexadec-9-enoyl]oxy]propyl (E)-hexadec-9-enoate
SMILES CCCCCC\C=C\CCCCCCCC(=O)OCC(COC(=O)CCCCCCC\C=C\CCCCCC)OC(=O)CCCCCCC\C=C\CCCCCC

Specifications

Quantity 1 g
InChI Formula InChI=1S/C51H92O6/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-49(52)55-46-48(57-51(54)45-42-39-36-33-30-27-24-21-18-15-12-9-6-3)47-56-50(53)44-41-38-35-32-29-26-23-20-17-14-11-8-5-2/h19-24,48H,4-18,25-47H2,1-3H3/b22-19+,23-20+,24-21+
Formula Weight 800.6894
Chemical Name or Material Glyceryl Tripalmitelaidate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.