CAS RN 134907-85-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Glyceryl Tripalmitelaidate, TRC
CAS: 134907-85-0 Molecular Formula: C51H92O6 Molecular Weight (g/mol): 801.27 Synonym: (9E,9'E,9''E)-9-Hexadecenoic Acid 1,2,3-Propanetriyl Ester,(E,E,E)-9-Hexadecenoic Acid 1,2,3-Propanetriyl Ester,Tripalmitelaidin IUPAC Name: 2,3-bis[[(E)-hexadec-9-enoyl]oxy]propyl (E)-hexadec-9-enoate SMILES: CCCCCC\C=C\CCCCCCCC(=O)OCC(COC(=O)CCCCCCC\C=C\CCCCCC)OC(=O)CCCCCCC\C=C\CCCCCC