missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Glutathione Sepharose™ 4B Media

Use for high capacity purification of Glutathione S-Transferase (GST) tagged fusion proteins. Cytiva Life Sciences™ Glutathione Sepharose™ 4B Media requires only a single-step for purification.

Brand:  Cytiva 17-0756-05

Code : GE

Additional Details : Weight : 1.36080kg


 View more versions of this product

Product Code. 11590794

  • £1539.00 / 100mL
Estimated Shipment: 15-04-2024
to see stock.
Add to basket

Description

Description

  • Binding capacity: >5mg horse liver GST/mL medium (flowrate dependent) Average particle size: 90μm
  • Recommended flowrate (H2O at room temp.): 75cm/hour
  • Stable in presence of all commonly used aqueous buffers
  • Stable in pH range 4 to 13

Specifications

Specifications

Glutathione Sepharose 4 B
4 to 13
90 μm
100 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Glutathione Sepharose 4B, 100 ml

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501-Dispose of contents/container to .

Supplemental information
  • MIXTURE LIST-Contains : Ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers