missing translation for 'onlineSavingsMsg'
Learn More

Glutamine Synthetase Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

£178.00 - £375.00

Specifications

Antigen Glutamine Synthetase
Applications Western Blot, Immunohistochemistry, Immunofluorescence, KnockDown
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18350734
View Documents
Novus Biologicals
NBP3-03471-20UL
20 μL
£178.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18372324
View Documents
Novus Biologicals
NBP3-03471-100UL
100 μL
£375.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Glutamine Synthetase Polyclonal specifically detects Glutamine Synthetase in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated.
Specifications

Specifications

Glutamine Synthetase
Polyclonal
Purified
RUO
Human, Mouse
cell proliferation-inducing protein 59, EC 4.1.1.15, EC 6.3.1.2, GLNSPIG59, Glutamate decarboxylase, glutamate-ammonia ligase, Glutamate--ammonia ligase, glutamate-ammonia ligase (glutamine synthase), glutamine synthase, glutamine synthetase, GS, PIG43, proliferation-inducing protein 43
Recombinant fusion protein containing a sequence corresponding to amino acids 1-373 of human Glutamine Synthetase (NP_001028228.1).
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot, Immunohistochemistry, Immunofluorescence, KnockDown
Unconjugated
Rabbit
Hypoxia, Neurodegeneration, Neuroscience, Vision
PBS with 50% glycerol, pH7.3.
2752
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.