missing translation for 'onlineSavingsMsg'
Learn More

Glutamine Synthetase Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Product Code. 18372324 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18372324 100 μL 100µL
18350734 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18372324 Supplier Novus Biologicals Supplier No. NBP303471100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Glutamine Synthetase Polyclonal specifically detects Glutamine Synthetase in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glutamine Synthetase
Applications Western Blot, Immunohistochemistry, Immunofluorescence, KnockDown
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias cell proliferation-inducing protein 59, EC 4.1.1.15, EC 6.3.1.2, GLNSPIG59, Glutamate decarboxylase, glutamate-ammonia ligase, Glutamate--ammonia ligase, glutamate-ammonia ligase (glutamine synthase), glutamine synthase, glutamine synthetase, GS, PIG43, proliferation-inducing protein 43
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-373 of human Glutamine Synthetase (NP_001028228.1).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Hypoxia, Neurodegeneration, Neuroscience, Vision
Primary or Secondary Primary
Gene ID (Entrez) 2752
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.