missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GLUT4 Recombinant Rabbit Monoclonal Antibody (7Y4R9)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£147.00 - £363.00

Specifications

Antigen GLUT4
Clone 7Y4R9
Concentration 1.69 mg/mL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Applications ELISA, Western Blot
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30283027
View Documents
Invitrogen™
MA558233
20 μL
£147.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30282698
GREENER_CHOICE
View Documents
Invitrogen™
MA558234
100 μL
£363.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Immunogen sequence: RPLSLLQLLG SRTHRQPLII AVVLQLSQQL SGINAVFYYS TSIFETAGVG QPAYATIGAG VVNTVFTLVS VLLVERAGRR TLHLLGLAGM CGCAILMTVA.

This gene is a member of the solute carrier family 2 (facilitated glucose transporter) family and encodes a protein that functions as an insulin-regulated facilitative glucose transporter. In the absence of insulin, this integral membrane protein is sequestered within the cells of muscle and adipose tissue. Within minutes of insulin stimulation, the protein moves to the cell surface and begins to transport glucose across the cell membrane. Mutations in this gene have been associated with noninsulin-dependent diabetes mellitus (NIDDM).
Specifications

Specifications

GLUT4
1.69 mg/mL
ELISA, Western Blot
Unconjugated
Rabbit
RUO
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
P14142
20528
Synthetic peptide.
Antibody
7Y4R9
-20°C, Avoid Freeze/Thaw Cycles
Recombinant Monoclonal
Liquid
IgG
Mouse
Slc2a4
glucose transporter 4; Glucose transporter 4 insuline-responsive; Glucose transporter 4, insuline-responsive; glucose transporter GLUT4; glucose transporter type 4; glucose transporter type 4, insulin-responsive; GLUT4; GLUT-4; GT2; insulin responsive glucose transporter; insulin-responsive glucose transporter; insulin-responsive glucose transporter type 4; MGC93607; SLC2A4; solute carrier family 2 member 4; solute carrier family 2 (facilitated glucose transporter), member 4; solute carrier family 2 , member 4; solute carrier family 2 member 4; solute carrier family 2, facilitated glucose transporter member 4
Slc2a4
Primary
Protein A
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.