missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GLUT4 Recombinant Rabbit Monoclonal Antibody (7Y4R9)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA558234

 View more versions of this product

Product Code. 30282698

  • £363.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: RPLSLLQLLG SRTHRQPLII AVVLQLSQQL SGINAVFYYS TSIFETAGVG QPAYATIGAG VVNTVFTLVS VLLVERAGRR TLHLLGLAGM CGCAILMTVA.

This gene is a member of the solute carrier family 2 (facilitated glucose transporter) family and encodes a protein that functions as an insulin-regulated facilitative glucose transporter. In the absence of insulin, this integral membrane protein is sequestered within the cells of muscle and adipose tissue. Within minutes of insulin stimulation, the protein moves to the cell surface and begins to transport glucose across the cell membrane. Mutations in this gene have been associated with noninsulin-dependent diabetes mellitus (NIDDM).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GLUT4
Recombinant Monoclonal
1.69 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
P14142
Slc2a4
Synthetic peptide.
100 μL
Primary
Mouse
Antibody
IgG
ELISA, Western Blot
7Y4R9
Unconjugated
Slc2a4
glucose transporter 4; Glucose transporter 4 insuline-responsive; Glucose transporter 4, insuline-responsive; glucose transporter GLUT4; glucose transporter type 4; glucose transporter type 4, insulin-responsive; GLUT4; GLUT-4; GT2; insulin responsive glucose transporter; insulin-responsive glucose transporter; insulin-responsive glucose transporter type 4; MGC93607; SLC2A4; solute carrier family 2 member 4; solute carrier family 2 (facilitated glucose transporter), member 4; solute carrier family 2 , member 4; solute carrier family 2 member 4; solute carrier family 2, facilitated glucose transporter member 4
Rabbit
Protein A
RUO
20528
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.